Skip to Content
Merck
All Photos(4)

Key Documents

A0781

Sigma-Aldrich

Aphidicolin from Nigrospora sphaerica

≥98% (HPLC), powder

Synonym(s):

APC

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$358.00
5 MG
$973.00
10 MG
$1,940.00

$358.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$358.00
5 MG
$973.00
10 MG
$1,940.00

About This Item

Empirical Formula (Hill Notation):
C20H34O4
CAS Number:
Molecular Weight:
338.48
Beilstein:
4689958
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$358.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]27/D +12°, c = 1 in methanol(lit.)

color

white

mp

227-233 °C (lit.)

solubility

ethanol: 1 mg/mL (stable at least a week at 4°C.)
DMSO: 10 mg/mL (stable at least six weeks at −20°C.)
methanol: 10 mg/mL
H2O: insoluble

antibiotic activity spectrum

neoplastics
viruses

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@@]3([H])[C@](C)(CO)[C@H](O)CC[C@]3(C)[C@]14CC[C@](O)(CO)[C@]([H])(C2)C4

InChI

1S/C20H34O4/c1-17(11-21)15-4-3-13-9-14-10-19(13,7-8-20(14,24)12-22)18(15,2)6-5-16(17)23/h13-16,21-24H,3-12H2,1-2H3/t13-,14+,15-,16+,17-,18-,19-,20-/m0/s1

InChI key

NOFOAYPPHIUXJR-APNQCZIXSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Aphidicolin is an antibiotic, a potent antiviral and antimitotic agent and DNA polymerase inhibitor.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Caution

Estimated shelf-life is two years. For optimal stability, it should be protected from light and stored desiccated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masao Takeuchi et al.
PloS one, 10(5), e0126562-e0126562 (2015-05-16)
Comprehensive analysis of alterations in gene expression along with neoplastic transformation in human cells provides valuable information about the molecular mechanisms underlying transformation. To further address these questions, we performed whole transcriptome analysis to the human mesenchymal stem cell line
Devin Sok et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(49), 17624-17629 (2014-11-26)
Broadly neutralizing antibodies (bnAbs) targeting the trimer apex of HIV envelope are favored candidates for vaccine design and immunotherapy because of their great neutralization breadth and potency. However, methods of isolating bnAbs against this site have been limited by the
Edwin Leeansyah et al.
PLoS pathogens, 11(8), e1005072-e1005072 (2015-08-22)
Mucosa-associated invariant T (MAIT) cells represent a large innate-like evolutionarily conserved antimicrobial T-cell subset in humans. MAIT cells recognize microbial riboflavin metabolites from a range of microbes presented by MR1 molecules. MAIT cells are impaired in several chronic diseases including
Athan C Baillet et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(6), 1535-1547 (2015-01-28)
Chlamydia trachomatis is a sexually transmitted obligate intracellular pathogen that causes inflammatory reactive arthritis, spondylitis, psoriasiform dermatitis, and conjunctivitis in some individuals after genital infection. The immunologic basis for this inflammatory response in susceptible hosts is poorly understood. As ZAP-70(W163C)
Julian H Elliott et al.
PLoS pathogens, 10(10), e1004473-e1004473 (2014-11-14)
Human immunodeficiency virus (HIV) persistence in latently infected resting memory CD4+ T-cells is the major barrier to HIV cure. Cellular histone deacetylases (HDACs) are important in maintaining HIV latency and histone deacetylase inhibitors (HDACi) may reverse latency by activating HIV

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service