Skip to Content
Merck
All Photos(1)

Key Documents

90423

Sigma-Aldrich

UV-VIS detectable IEF Marker solution

pI 6.6, 1 mg/mL in 10 mM NaOH, 0.45 μm filtered

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$339.00

$339.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
200 μL
$339.00

About This Item

UNSPSC Code:
41105336
NACRES:
NA.26

$339.00


Estimated to ship on02 June 2025


Request a Bulk Order

sterility

0.45 μm filtered

form

solution

concentration

1 mg/mL in 10 mM NaOH

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Suitable as IEF marker for HPCE and IEF gels.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service