82452
Proteinase K, immobilized on Eupergit® C from Tritirachium album
powder (granular), ≥1500 U/g
Synonym(s):
Endopeptidase K, immobilized on Eupergit® C
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
NACRES:
NA.54
Recommended Products
biological source
fungus (Tritirachium album)
form
powder (granular)
specific activity
≥1500 U/g
storage temp.
2-8°C
General description
Proteinase K is secreted extracellular medium by mold, Tritirachium album. It hydrolyses keratin and shows substrate specificity similar to serine alkaline proteinases.
Application
Proteinase K, immobilized on Eupergit® C from Tritirachium album has been used:
- in the protolysis of human serum
- in the digestion of gruel samples for enzyme linked immune assays (ELISA)
- in proteolytic stability studies of calcium ion flux inducers
Biochem/physiol Actions
Proteinase K hydrolyses keratin by cleaving peptide bonds adjacent to the carboxyl group of aliphatic and aromatic amino acids.
Unit Definition
1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol N-acetyl-L-tyrosine-ethylester (ATEE) per minute at pH 9.0 and 30 °C.
Legal Information
Eupergit is a registered trademark of Röhm GmbH & Co. KG
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Specificity of proteinase K from Tritirachium album Limber for synthetic peptides
MoriharaKand Tsuzuki, Hiroshige
Agricultural and Biological Chemistry, 39(7), 1489-1492 (1975)
Enterococcus faecium stimulates human neutrophils via the formyl-peptide receptor 2
Bloes DA, et al.
Testing, 7(6), e39910-e39910 (2012)
Neil T Sprenkle et al.
Journal of neurochemistry, 148(4), 516-530 (2018-12-07)
Improper protein folding and trafficking are common pathological events in neurodegenerative diseases that result in the toxic accumulation of misfolded proteins within the lumen of the endoplasmic reticulum (ER). While low-level stimulation of the unfolded protein response (UPR) is protective
Isotope dilution ESI-LC-MS/MS for quantification of free and total Nvarepsilon-(1-Carboxymethyl)-l-Lysine and free Nvarepsilon-(1-Carboxyethyl)-l-Lysine: Comparison of total Nvarepsilon-(1-Carboxymethyl)-l-Lysine levels measured with new method to ELISA assay in gruel samples
Tareke E, et al.
Food Chemistry, 141(4), 4253-4259 (2013)
Lysophosphatidic acid cooperates with 1alpha, 25 (OH) 2D3 in stimulating human MG63 osteoblast maturation
Gidley J, et al.
Prostaglandins & other lipid mediators, 80(1-2), 46-61 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service