Skip to Content
Merck
All Photos(1)

Key Documents

803057

Sigma-Aldrich

Manganese Peroxidase from white-rot fungus (Phanerochaete chrysosporium)

recombinant, expressed in corn, lyophilized powder, ≥200 units/g solid

Synonym(s):

Manganese peroxidase from white-rot fungus (Phanerochaete chrysosporium), Manganese-dependent lignin peroxidase, MnP, Peroxidase, manganese

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$615.00

$615.00


Check Cart for Availability


Select a Size

Change View
10 MG
$615.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

$615.00


Check Cart for Availability

recombinant

expressed in corn

Quality Level

form

lyophilized powder

specific activity

≥200 units/g solid

mol wt

~53 kDa (single isozyme)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C (−15°C to −25°C)

Looking for similar products? Visit Product Comparison Guide

General description

Manganese peroxidase from white-rot fungus (Phanerochaete chrysosporium) is a lignin-modifying peroxidase that catalyzes the oxidation of Mn2+ to Mn3+. Mn3+ is stabilized by chelators like oxalic acid. Manganese peroxidase uses peroxide as an oxidant and manganese coordination compounds as charge-transfer mediators.

Manganese peroxidase has a molecular weight of 40-50 kDa and is a glycosylated heme protein.[1][2]

Unit Definition

One unit is the amount of enzyme which will oxidize 1.0 μmole Mn2+ per minute to Mn3+ at pH 4.5 and 25 °C

Physical form

Supplied as a lyophilized powder containing ~50% sodium tartrate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Review: lignin conversion by manganese peroxidase (MnP)
Martin H
Enzyme and Microbial Technology, 30(4) (2002)
Kinetic Analysis of Manganese Peroxidase
I-Ching Kuan
The Journal of Biological Chemistry, 268 (1993)

Questions

1–2 of 2 Questions  
  1. Do you have a protocol that you recommend for the analysis of manganese peroxidase activity?

    1 answer
    1. The Technical Service team can provide the requested documentation for the analysis of manganese peroxidase activity. Please visit https://www.sigmaaldrich.com/techservice and click on "Product Documentation" under the Products Section to submit the required information for further assistance.

      Helpful?

  2. 해당 효소의 유전자 서열과 균주의 strain을 알고 싶습니다

    1 answer
    1. The gene sequence is considered proprietary. The species is Phanerochaete chrysosporium. This is a recombinant protein expressed in corn.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service