
Manganese Peroxidase from white-rot fungus (Phanerochaete chrysosporium)

Synonym(s):
Manganese peroxidase from white-rot fungus (Phanerochaete chrysosporium), Manganese-dependent lignin peroxidase, MnP, Peroxidase, manganese
CAS Number:
Products
Product Category
Brand
Physical Form
Recombinant Host
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing