Skip to Content
Merck
All Photos(1)

Key Documents

75452

Sigma-Aldrich

2,6-Diethyl-4,4-difluoro-1,3,5,7-tetramethyl-8-[4-(2-propinyloxy)phenyl]-4-bora-3a,4a-diaza-s-indacene

for click labeling

Synonym(s):

Bodipy-X-Alkyne

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29BF2N2O
CAS Number:
Molecular Weight:
434.33
Beilstein:
19893171
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

form

solid

solubility

methanol: soluble, clear, orange (to very dark orange)

fluorescence

λex 525 nm; λem 535 nm±5 nm in methanol

storage temp.

−20°C

SMILES string

CCc1c(C)c2C(c3ccc(OCC#C)cc3)=C4C(C)=C(CC)C(C)=[N+]4[B-](F)(F)n2c1C

InChI

1S/C26H29BF2N2O/c1-8-15-32-21-13-11-20(12-14-21)24-25-16(4)22(9-2)18(6)30(25)27(28,29)31-19(7)23(10-3)17(5)26(24)31/h1,11-14H,9-10,15H2,2-7H3

InChI key

BAUOLRCAARTXSE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service