Skip to Content
Merck
All Photos(1)

Key Documents

901214

Sigma-Aldrich

1,1′-[4,8-Bis[5-(2-ethylhexyl)-2-thienyl]benzo[1,2-b:4,5-b′]dithiophene-2,6-diyl]bis[1,1,1-trimethylstannane]

Synonym(s):

2,6-Bis(trimethylstannyl)-4,8-bis(5-(2-ethylhexyl)thiophen-2-yl)benzo[1,2-b:4,5-b′]dithiophene, 2,6-Bis(trimethyltin)-4,8-bis[5-(2-ethylhexyl)thiophen-2-yl]benzo[1,2-b:4,5-b′]dithiophene

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$521.00

$521.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$521.00

About This Item

Empirical Formula (Hill Notation):
C40H58S4Sn2
CAS Number:
Molecular Weight:
904.57
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.23

$521.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

Quality Level

form

powder

storage temp.

2-8°C

SMILES string

[Sn](C)(C)(C)c1[s]c2c(c(c4[s]c(cc4c2c5[s]c(cc5)CC(CCCC)CC)[Sn](C)(C)C)c3[s]c(cc3)CC(CCCC)CC)c1

InChI

1S/C34H40S4.6CH3.2Sn/c1-5-9-11-23(7-3)21-25-13-15-29(37-25)31-27-17-19-36-34(27)32(28-18-20-35-33(28)31)30-16-14-26(38-30)22-24(8-4)12-10-6-2;;;;;;;;/h13-18,23-24H,5-12,21-22H2,1-4H3;6*1H3;;

InChI key

OCFFMJYHZKHRKM-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haijun Bin et al.
Journal of the American Chemical Society, 139(14), 5085-5094 (2017-03-23)
In the last two years, polymer solar cells (PSCs) developed quickly with n-type organic semiconductor (n-OSs) as acceptor. In contrast, the research progress of nonfullerene organic solar cells (OSCs) with organic small molecule as donor and the n-OS as acceptor
Liyan Yang et al.
Journal of the American Chemical Society, 139(5), 1958-1966 (2017-01-14)
A new organic small molecule, DRTB-T, that incorporates a two-dimensional trialkylthienyl-substituted benzodithiophene core building block was designed and synthesized. DRTB-T has a band gap (E

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service