74134
epi-Inositol
≥98.0% (HPLC)
Synonym(s):
epi-Cyclohexanehexol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥98.0% (HPLC)
form
crystals
storage temp.
2-8°C
SMILES string
O[C@@H]1[C@@H](O)[C@H](O)[C@H](O)[C@H](O)[C@@H]1O
InChI
1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2-,3-,4+,5-,6+
InChI key
CDAISMWEOUEBRE-NIPYSYMMSA-N
Biochem/physiol Actions
epi-Inositol has regulatory effects on expression of the most highly regulated gene in the inositol biosynthetic pathway.
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kieran P Stockton et al.
The Journal of organic chemistry, 79(11), 5088-5096 (2014-04-23)
A synthesis of carbocyclic sugars from carbohydrate-derived dialdehydes using organocatalysis has been developed. Sorbitol, mannitol, and galactitol were converted via 1,6-tritylation, perbenzylation or permethylation, detritylation, and Swern oxidation into 2,3,4,5-tetra-O-alkyl-dialdoses that were cyclized via the benzoin reaction promoted by a
JoAnne McLaurin et al.
Nature medicine, 12(7), 801-808 (2006-06-13)
When given orally to a transgenic mouse model of Alzheimer disease, cyclohexanehexol stereoisomers inhibit aggregation of amyloid beta peptide (Abeta) into high-molecular-weight oligomers in the brain and ameliorate several Alzheimer disease-like phenotypes in these mice, including impaired cognition, altered synaptic
A Shaldubina et al.
Molecular psychiatry, 7(2), 174-180 (2002-02-13)
Myo-inositol exerts behavioral effects in animal models of psychiatric disorders and is effective in clinical trials in psychiatric patients. Interestingly, epi-inositol exerts behavioral effects similar to myo-inositol, even though epi-inositol is not a substrate for synthesis of phosphatidylinositol. We postulated
J McLaurin et al.
The Journal of biological chemistry, 275(24), 18495-18502 (2000-04-15)
Inositol has 8 stereoisomers, four of which are physiologically active. myo-Inositol is the most abundant isomer in the brain and more recently shown that epi- and scyllo-inositol are also present. myo-Inositol complexes with Abeta42 in vitro to form a small
Xiao-Long Liu et al.
Pesticide biochemistry and physiology, 164, 7-13 (2020-04-15)
Insect gustatory system plays important roles in multiple behaviors including feeding, mating, and oviposition. Gustatory receptors (GRs), located on the dendritic membrane of gustatory sensory neurons (GSNs), are crucial in peripheral coding of non-volatile compounds. However, GRs and their detailed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service