Skip to Content
Merck
All Photos(1)

Documents

51445C

SAFC

Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F12

liquid, suitable for cell culture

Synonym(s):

Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham, DME/F-12, 1:1 mixture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use
with 3151 mg/L dextrose, with 2.5 mM L-glutamine, with 15mM HEPES, with 55 mg/L sodium pyruvate

sterility

sterile

form

liquid

technique(s)

cell culture | mammalian: suitable

components

sodium pyruvate: 0.055 g/L
phenol red: yes
glucose: 3.15 g/L
L-glutamine: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

DMEM/F12 was originally developed to study hormonal requirements of cells in culture.

Application

Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F12 has been used as a culture media for mouse muscle myoblast C2C12 and human bone marrow cell line SH-SY5Y.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

hmSOD1 gene mutation?induced disturbance in iron metabolism is mediated by impairment of Akt signalling pathway.
Halon?Golabek M, et al.
Journal of Cachexia, Sarcopenia and Muscle (2018)
Yubin Pan et al.
In vitro cellular & developmental biology. Animal, 56(2), 129-144 (2020-01-17)
During hematogenous metastasis, cancer cells escape from primary lesions and enter into the circulatory system, and only a few can colonize distant organs. However, the mechanism of cell survival and metastasis in the hematopoietic environment remains unclear. Angiorrhea is the
Malgorzata Halon-Golabek et al.
Journal of cachexia, sarcopenia and muscle, 9(3), 557-569 (2018-01-31)
Recently, skeletal muscle atrophy, impairment of iron metabolism, and insulin signalling have been reported in rats suffering from amyotrophic lateral sclerosis (ALS). However, the interrelationship between these changes has not been studied. We hypothesize that an impaired Akt-FOXO3a signalling pathway
Yan Xing et al.
Biology open, 9(4) (2020-03-07)
Infants with intrauterine growth retardation (IUGR) have a high risk of developing bronchial asthma in childhood, but the underlying mechanisms remain unclear. This study aimed to disclose the role of vascular non-inflammatory molecule 1 (vannin-1, encoded by the Vnn1 gene)
Dominika Czerwonka et al.
ChemMedChem, 15(2), 236-246 (2019-11-09)
The polyether ionophore salinomycin (SAL) has captured much interest because of its potent activity against cancer cells and cancer stem cells. Our previous studies have indicated that C1/C20 double-modification of SAL is a useful strategy to generate diverse agents with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service