Skip to Content
Merck
All Photos(1)

Key Documents

56498C

SAFC

Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F12

powder, with 3151 mg/L dextrose, with 2.5 mM L-glutamine, with 55 mg/L sodium pyruvate, without sodium bicarbonate

Pharma Manufacturing

Synonym(s):

Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham, DME/F-12, 1:1 mixture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.71

description

for research or for further manufacturing use

Quality Level

form

powder

components

phenol red: yes
L-glutamine: yes
sodium pyruvate: 0.055 g/L
glucose: 3.15 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Dulbecco′s Modified Eagle′s medium (DMEM) in combination with Ham′s Nutrient Mixture F12 has been developed to study the hormonal requirements of cells in culture.

Application

Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F12 has been used to culture:
  • osteosarcoma cells
  • mouse microglial cell line
  • human gastric adenocarcinoma cell lines

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vikas Singh et al.
Bio-protocol, 7(11), e2304-e2304 (2017-06-05)
Glutathione is one of the major antioxidant defense components present in cells. It is predominantly present as reduced glutathione (GSH) and converted into oxidized glutathione (GSSG) while reducing the free radicals like hydroxyl ions (OH-). For the measurement of GSH
Kavitha Kandiah et al.
Materials science & engineering. C, Materials for biological applications, 38, 252-262 (2014-03-25)
Bone defects and damages are common these days, which increases the usage of biomaterial for humans. To prepare a potential biomaterial, we synthesised a series of titania-graphene nanocomposites (TGS) (2:x (0.25, 0.5, 1.0, 2.0, and 4.0 g)) using in situ
Yanyan Miao et al.
Biomaterials, 149, 29-40 (2017-10-11)
Cancer stem cells (CSCs) are believed to have a critical role in tumorigenesis, metastasis, therapeutic resistance or recurrence. Therefore, strategies designed to specifically target and eliminate CSCs have become one of the most promising and desirable ways for tumor treatment.
Vikas Singh et al.
Bio-protocol, 7(11), e2304-e2304 (2017-06-05)
Glutathione is one of the major antioxidant defense components present in cells. It is predominantly present as reduced glutathione (GSH) and converted into oxidized glutathione (GSSG) while reducing the free radicals like hydroxyl ions (OH-). For the measurement of GSH
Tobias M Lindig et al.
Histochemistry and cell biology, 131(1), 1-12 (2008-09-23)
We have compared the three-dimensional (3D) morphology of stubby and spiny neurons derived from the human small intestine. After immunohistochemical triple staining for leu-enkephalin (ENK), vasoactive intestinal peptide (VIP) and neurofilament (NF), neurons were selected and scanned based on their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service