Skip to Content
Merck
All Photos(1)

Key Documents

4427700

Sigma-Aldrich

mTRAQ® Reagent Δ8 50 Unit Pack

Synonym(s):

Δ8 mTRAQ Reagent Pack, 50 Unit mTRAQ Reagent

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$5,280.00

$5,280.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$5,280.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

$5,280.00


Check Cart for Availability

Request a Bulk Order

storage temp.

−20°C

General description

Contains mTRAQ® Reagents Δ8 for 50 mTRAQ assays. One unit of reagent can label up to 100 μg of protein. Pack has 1 tube each of Δ8 containing 50 units of reagent. Optional Multiplex Buffer kit (Cat. No. 4381664) to be ordered separately.

Analysis Note

To view the Protocol for use of mTRAQ Reagents, please visit this Protocol link.

To view the Quick Reference Card for use of mTRAQ Reagents, please visit this Quick Reference Card link.

Legal Information

MTraq is a registered trademark of AB Sciex Pte. Ltd.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service