Skip to Content
Merck
All Photos(1)

Documents

72723

Supelco

Fucoxanthinol

analytical standard

Synonym(s):

all-trans-Fucoxanthinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56O5
CAS Number:
Molecular Weight:
616.87
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

−20°C

SMILES string

C/C(C=C=C1C(C)(C)C[C@H](O)C[C@]1(O)C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(C)\C(C[C@@]2(O3)[C@@]3(C)C[C@@H](O)CC2(C)C)=O

InChI

1S/C40H56O5/c1-28(17-13-18-30(3)21-22-35-36(5,6)23-32(41)25-38(35,9)44)15-11-12-16-29(2)19-14-20-31(4)34(43)27-40-37(7,8)24-33(42)26-39(40,10)45-40/h11-21,32-33,41-42,44H,23-27H2,1-10H3/b12-11+,17-13+,19-14+,28-15+,29-16+,30-18+,31-20+/t22-,32-,33-,38+,39+,40-/m0/s1

InChI key

NZEPSBGUXWWWSI-VOPPYBPXSA-N

General description

Fucoxanthinol belongs to the class of carotenoids. It is the primary metabolite of fucoxanthin, a major marine carotenoid isolated from edible brown seaweeds such as Undaria pinnatifida, Hijikia fusiformis, Laminaria japonica and Sargassum fulvellum with various biological functions. Fucoxanthinol is a compound of significant interest as it exhibits various physiological effects. It is a chemotherapeutic agent that is known to inhibit the growth of human colorectal cancer cells.

Application

Fucoxanthinol may be used as an analytical reference standard for the determination of the analyte in rat plasma by chromatography-based techniques.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of fucoxanthin and its deacetylated metabolite fucoxanthinol in rat plasma by liquid chromatography-tandem mass spectrometry
Zhang , et al.
Marine drugs, 13(10), 6521- 6536 (2015)
Fucoxanthin and its metabolite, fucoxanthinol, suppress adipocyte differentiation in 3T3-L1 cells
Maeda H, et al.
International Journal of Molecular Medicine, 18(1), 147- 152 (2006)
Determination of fucoxanthinol in rat plasma by liquid chromatography-tandem mass spectrometry
Zhang Y, et al.
Journal of Pharmaceutical and Biomedical Analysis, 164(3), 155- 163 (2019)
Anticancer effects of fucoxanthin and fucoxanthinol on colorectal cancer cell lines and colorectal cancer tissues
Takahashi K, et al.
Oncology Letters, 10(3), 1463- 1467 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service