Skip to Content
Merck
All Photos(1)

Key Documents

909025

Sigma-Aldrich

Lithium hydroxide

free-flowing, Redi-Dri, powder, reagent grade, ≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$172.00
250 G
$252.00

$172.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$172.00
250 G
$252.00

About This Item

Linear Formula:
LiOH
CAS Number:
Molecular Weight:
23.95
MDL number:
UNSPSC Code:
12352305
NACRES:
NA.21
grade:
reagent grade
form:
powder

$172.00


Check Cart for Availability

Request a Bulk Order

grade

reagent grade

product line

Redi-Dri

Assay

≥98%

form

powder

mp

470 °C (dec.) (lit.)

SMILES string

[Li+].[OH-]

InChI

1S/Li.H2O/h;1H2/q+1;/p-1

InChI key

WMFOQBRAJBCJND-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Redi-Dri is an innovative packaging product line that helps eliminate clumping material by adding extra packaging process steps without any chemical additives, anti-clumping agents, or hydrophobic compounds. Redi Dri offers a more convenient and safer way to handle the products.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service