Skip to Content
Merck
All Photos(3)

Key Documents

545856

Sigma-Aldrich

Lithium hydroxide

Lithium hydroxide

powder, reagent grade, ≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$121.00
500 G
$495.00

$121.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$121.00
500 G
$495.00

About This Item

Linear Formula:
LiOH
CAS Number:
Molecular Weight:
23.95
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
form:
powder
solubility:
water: soluble 71 g/L at 20 °C

$121.00


Available to ship on14 April 2025Details


Request a Bulk Order

grade

reagent grade

Assay

≥98%

form

powder

mp

470 °C (dec.) (lit.)

solubility

water: soluble 71 g/L at 20 °C

SMILES string

[Li+].[OH-]

InChI

1S/Li.H2O/h;1H2/q+1;/p-1

InChI key

WMFOQBRAJBCJND-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium hydroxide (LiOH) is an alkali metal hydroxide.[1] Lithium chloride solution in water on electrolysis forms LiOH. In respiratory apparatus and submarines, it is utilized to uptake carbon dioxide.[2] A study on the redox mechanism of titanium dioxide (TiO2) using cyclic voltammetry, X-ray diffraction, X-ray photoelectron spectroscopy (XPS) and Fourier transform infrared spectroscopy (FTIR) in aqueous LiOH electrolyte has been reported.[3]

Application

Lithium hydroxide has been used in the following processes:
  • Synthesis of lithium-doped zinc oxide (ZnO) thin films.[1]
  • Preparation of lithium glyceroxide/hydroxide catalysts by reacting with glycerol.[4]
  • As a catalyst to generate unsaturated ketones via Michael addition of β-dicarbonyl compounds.[5]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 605-605 (1994)
Lithium Hydroxide-Catalyzed Conjugate Addition of β-Dicarbonyl Compounds.
Bonadies F, et al.
ChemInform, 26(23) (1995)
Production of biodiesel with lithium glyceroxide.
Wang E, et al.
Fuel: The Science and Technology of Fuel and Energy, 160, 621-628 (2015)
Low-Temperature, Solution-Processed and Alkali Metal Doped ZnO for High-Performance Thin-Film Transistors.
Park SY, et al.
Advanced Mat., 24(6), 834-838 (2012)
Electrochemical behavior of anatase TiO2 in aqueous lithium hydroxide electrolyte.
Manickam M, et al.
J. Appl. Electrochem., 36(5), 599-602 (2006)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service