Skip to Content
Merck
All Photos(2)

Documents

675326

Sigma-Aldrich

Activated charcoal

greener alternative

DARCO® KB-G

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52

vapor pressure

<0.1 mmHg ( 20 °C)

form

powder

autoignition temp.

842 °F

manufacturer/tradename

DARCO® KB-G

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

resistivity

1375 μΩ-cm, 20°C (graphite)

impurities

≤15.0% moisture content

particle size

>45 μm

mp

3550 °C (lit.)

anion traces

phosphate (PO43-): ≤3.5%

cation traces

Ca: ≤200 ppm
Fe: ≤150 ppm

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Suitable for the purification of highly colored and/or foaming process liquids.

Legal Information

DARCO is a registered trademark of Cabot Norit Americas, Inc.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kohsuke Ohmatsu et al.
Nature chemistry, 6(1), 47-51 (2013-12-19)
The development of a general catalytic method for the direct and stereoselective construction of contiguous all-carbon quaternary stereocentres remains a formidable challenge in chemical synthesis. Here, we report a highly enantio- and diastereoselective [3+2] annulation reaction of 5-vinyloxazolidinones and activated
Nathalie Butt et al.
Ecological applications : a publication of the Ecological Society of America, 23(4), 936-943 (2013-07-20)
A typical way to quantify aboveground carbon in forests is to measure tree diameters and use species-specific allometric equations to estimate biomass and carbon stocks. Using "citizen scientists" to collect data that are usually time-consuming and labor-intensive can play a
Yin Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(1), 533-538 (2013-12-25)
Stomatal pores surrounded by a pair of guard cells in the plant epidermis control gas exchange between plants and the atmosphere in response to light, CO2, and the plant hormone abscisic acid. Light-induced stomatal opening is mediated by at least
Nuno Carvalhais et al.
Nature, 514(7521), 213-217 (2014-09-26)
The response of the terrestrial carbon cycle to climate change is among the largest uncertainties affecting future climate change projections. The feedback between the terrestrial carbon cycle and climate is partly determined by changes in the turnover time of carbon
Michael C Dietze et al.
Annual review of plant biology, 65, 667-687 (2013-11-28)
Nonstructural carbon (NSC) provides the carbon and energy for plant growth and survival. In woody plants, fundamental questions about NSC remain unresolved: Is NSC storage an active or passive process? Do older NSC reserves remain accessible to the plant? How

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service