Skip to Content
Merck
All Photos(2)

Key Documents

47670

Sigma-Aldrich

Formamide

puriss. p.a., ACS reagent, ≥99.5% (GC/T)

Synonym(s):

Amide C1, Formic amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Beilstein:
505995
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
technique(s):
PCR: suitable
bp:
210 °C (lit.)
vapor pressure:
0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

vapor density

1.55 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

Assay

≥99.5% (GC/T)

form

liquid

autoignition temp.

932 °F

expl. lim.

2.7-19 %

technique(s)

PCR: suitable

impurities

≤0.02% free acid (as HCOOH)
≤0.1% water

refractive index

n20/D 1.447 (lit.)
n20/D 1.447

pH

4-10 (20 °C, 200 g/L)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

transition temp

transition temp 2.0-3.0 °C

density

1.132 g/mL at 20 °C
1.134 g/mL at 25 °C (lit.)

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤1 mg/kg

suitability

in accordance for miscibility

SMILES string

NC=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)

InChI key

ZHNUHDYFZUAESO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Formamide is a colorless organic solvent.[1] It has been reported to increase the precision of PCR (Polymerase Chain Reaction) studies.[2] Microwave spectral study of its isotopic forms suggests that the molecule of formamide is nonplanar in which the H2N-C group exhibits a shallow pyramid like structure.[3]

Application

Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Solubilization in formamide protects RNA from degradation.
Chomczynski P.
Nucleic Acids Research, 20(14), 3791-3791 (1992)
Microwave spectrum and molecular structure of formamide.
Costain CC and Dowling JM.
J. Chem. Phys. , 32(1), 158-165 (1960)
J Casey et al.
Nucleic acids research, 4(5), 1539-1552 (1977-01-01)
The thermal stabilities of RNA:DNA hybrids are substantially greater than those of DNA:DNA duplexes in aqueous electrolyte solutions containing high concentrations of formamide. Association rates to form DNA:DNA duplexes and DNA:RNA hybrids have been measured in these solvents. There is
The formamide method for the extraction of polysaccharides from haemolytic streptococci.
Fuller AT.
British Journal of Experimental Pathology, 32(1), 130-130 (1960)
Mado Vandewoestyne et al.
International journal of legal medicine, 123(5), 441-447 (2009-03-26)
Laser microdissection is a valuable tool for isolating specific cells from mixtures, such as male cells in a mixture with female cells, e.g., in cases of sexual assault. These cells can be stained with Y-chromosome-specific probes. We developed an automatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service