Merck
  • Rates of formation and thermal stabilities of RNA:DNA and DNA:DNA duplexes at high concentrations of formamide.
Rates of formation and thermal stabilities of RNA:DNA and DNA:DNA duplexes at high concentrations of formamide.
Nucleic acids research (1977-01-01)
J Casey, N Davidson
PMID19730
ABSTRACT
The thermal stabilities of RNA:DNA hybrids are substantially greater than those of DNA:DNA duplexes in aqueous electrolyte solutions containing high concentrations of formamide. Association rates to form DNA:DNA duplexes and DNA:RNA hybrids have been measured in these solvents. There is a temperature range in which DNA:DNA rates are negligible and RNA:DNA rates close to optimal.
Sigma-Aldrich
Formamide, puriss. p.a., ACS reagent, ≥99.5% (GC/T)
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Formamide, puriss. p.a., ACS reagent, ≥99.5% (GC/T)