Skip to Content
Merck
All Photos(1)

Key Documents

45960

Sigma-Aldrich

Isopropyl acetate

puriss. p.a., ≥99.5% (GC)

Synonym(s):

Acetic acid isopropyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$62.70
1 L
$181.00

$62.70


Available to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 ML
$62.70
1 L
$181.00

About This Item

Linear Formula:
CH3COOCH(CH3)2
CAS Number:
Molecular Weight:
102.13
Beilstein:
1740761
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.01
technique(s):
GC/GC: suitable
bp:
85-91 °C (lit.)
vapor pressure:
47 mmHg ( 20 °C)

$62.70


Available to ship on10 April 2025Details


Request a Bulk Order

vapor density

3.5 (vs air)

vapor pressure

47 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

894 °F

expl. lim.

1.8 %, 37 °F
8 %

technique(s)

GC/GC: suitable

impurities

≤0.005% free acetic acid
≤0.05% water

evapn. residue

≤0.002%

refractive index

n20/D 1.377 (lit.)
n20/D 1.377

bp

85-91 °C (lit.)

mp

−73 °C (lit.)

density

0.872 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CC(C)OC(C)=O

InChI

1S/C5H10O2/c1-4(2)7-5(3)6/h4H,1-3H3

InChI key

JMMWKPVZQRWMSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopropyl acetate is an isopropyl ester of acetic acid. It participates in the mesoporous Al-MCM-41 (Si/Al = 55 and 104) and Al, Zn-MCM-41 (Si/(Al+Zn)=52) molecular sieves catalyzed alkylation of m-cresol.[1] It is widely used for the incorporation of aroma to various cosmetics and food products. Vapor-liquid equilibria for its binary mixture with CO2 at higher pressures has been evaluated.[2]

Application

Isopropyl acetate may be employed as a model oxygenate compound to evaluate the catalytic efficiency of La0.8Sr0.2MnO3+x perovskite catalyst for the oxidation of various oxy-derivative compounds.[3] It may be used as extracting reagent for the N,N-dimethyl-2-[5-(cyanomethyl)-1H-indol-3-yl]ethylamine.[4]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved Fischer indole reaction for the preparation of N,N-dimethyltryptamines: Synthesis of L-695,894, a potent 5-HT1D receptor agonist.
Chen CY, et al.
The Journal of Organic Chemistry, 59(13), 3738-3741 (1994)
General study of catalytic oxidation of various VOCs over La0.8Sr0.2MnO3+x perovskite catalyst-influence of mixture.
Blasin-Aube V, et al.
Applied Catalysis. B, Environmental, 43(2), 175-186 (2003)
Vapor-liquid equilibria of carbon dioxide with isopropyl acetate, diethyl carbonate and ethyl butyrate at elevated pressures.
Cheng CH and Chen YP.
Fluid Phase Equilibria, 234(1), 77-83 (2005)
A novel route to produce thymol by vapor phase reaction of m-cresol with isopropyl acetate over Al-MCM-41 molecular sieves.
Shanmugapriya K, et al.
J. Catal., 224(2), 347-357 (2004)
R Akimoto et al.
The Journal of surgical research, 53(6), 572-577 (1992-12-01)
Methyl tert-butyl ether (MTBE) is an organic solvent that has been used to dissolve gallstones via a percutaneous transhepatic catheter into the gallbladder. To test whether MTBE might cause serious tissue injury if accidentally infused outside the gallbladder, the effect

Articles

Analysis of carbonate solvents in lithium-ion batteries like ethylene carbonate, dimethyl carbonate, diethyl carbonate, and more using SPB®-624 column.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service