Skip to Content
Merck
All Photos(1)

Key Documents

58997

Supelco

SUPELCOSIL LC-SCX (5 µm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Synonym(s):

SCX column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
L × I.D.:
25 cm × 4.6 mm
surface area:
170 m2/g
particle size:
5 μm
matrix active group:
sulfonic acid, propyl- phase
pore size:
120 Å
matrix:
silica gel, spherical particle platform

Product Name

SUPELCOSIL LC-SCX HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

Agency

suitable for USP L52

Quality Level

manufacturer/tradename

SUPELCOSIL

parameter

≤70 °C temp. limit
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

170 m2/g

matrix

silica gel, spherical particle platform

matrix active group

sulfonic acid, propyl- phase

particle size

5 μm

pore size

120 Å

pH range

2-7.5

application(s)

food and beverages

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

The LC-SCX cation-exchange columns have strongly acidic propylsulfonic acid groups and are used for separating cations. Adjust pH, ionic strength, and organic modifier concentration to control retention and selectivity.

Application

Supelcosil LC-SCX HPLC column was used during the purification and characterization of 8-hydroxyguanine (oh8Gua), from Human Urine. It was used as a cation exchange column, during measurement and extraction of water-soluble arsenic, freeze-dried marine animal tissues, using high performance liquid chromatography-inductively coupled plasma mass spectrometry (HPLC-ICP-MS). It was also used in the chromatographic separation of arsenic compounds.
Supelcosil LC-SCX HPLC column was used during the purification and characterization of 8-hydroxyguanine (oh8Gua), from Human Urine. It was used as a cation exchange column, during measurement and extraction of water-soluble arsenic, freeze-dried marine animal tissues, using high performance liquid chromatography-inductively coupled plasma mass spectrometry (HPLC-ICP-MS). It was also used in the chromatographic separation of arsenic compounds. It may be used as a cation-exchange column for the separation, identification and determination of arsenic compounds in oyster tissue and marine brown alga like Hijiki fuziforme using high performance liquid chromatography coupled with inductively coupled plasma mass spectrometry (HPLC-ICP-MS).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Measurement of water-soluble arsenic species in freeze-dried marine animal tissues by microwave-assisted extraction and HPLC-ICP-MS.
Kirby J and Maher W.
Journal of Analytical Atomic Spectrometry, 17 (8), 838-843 (2002)
Comparison of three methods for the extraction of arsenic compounds from the NRCC standard reference material DORM-2 and the brown alga Hijiki fuziforme
Kuehnelt D, et al.
Applied Organometallic Chemistry, 15(6), 445-456 (2001)
Urinary 8-hydroxy-2'-deoxyguanosine as a biological marker of in vivo oxidative DNA damage.
Shigenaga MK
Proceedings of the National Academy of Sciences of the USA, 86 (24), 9697-9701 (1989)
Determination of arsenic compounds in earthworms.
GEISZINGER A,.
Environmental Science & Technology, 32 (15), 2238-2243 (1998)
Determination of total cationic and total anionic arsenic species in oyster tissue using microwave-assisted extraction followed by HPLC?ICP-MS
Chatterjee A
Talanta, 51(2), 303-314 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service