Skip to Content
Merck
All Photos(2)

Key Documents

227668

Sigma-Aldrich

Magnesium carbonate hydroxide hydrate

99%

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$86.60

$86.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$86.60

About This Item

Empirical Formula (Hill Notation):
C4Mg4O12 · H2MgO2 · xH2O
Molecular Weight:
395.58 (anhydrous basis)
EC Number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.55
form:
powder

$86.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

powder

SMILES string

O.[Mg++].[Mg++].[Mg++].[Mg++].O[Mg]O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The origin of the exothermic peak in the thermal decomposition of basic magnesium carbonate.
Khan N, et al.
Thermochimica Acta, 367, 321-333 (2001)
Conazole fungicides inhibit Leydig cell testosterone secretion and androgen receptor activation in vitro.
Roelofs MJE, et al.
Toxicology Reports, 1, 271-283 (2014)
Preparation of a magnesium hydroxy carbonate from magnesium hydroxide.
Botha A and Strydom CA.
Hydrometallurgy, 62(3), 175-183 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service