Skip to Content
Merck
All Photos(5)

Key Documents

237175

Sigma-Aldrich

Magnesium nitrate hexahydrate

ACS reagent, 99%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$75.30
500 G
$109.00
2.5 KG
$436.00
1 KG
$467.00

$75.30


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 G
$75.30
500 G
$109.00
2.5 KG
$436.00
1 KG
$467.00

About This Item

Linear Formula:
Mg(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
256.41
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
98.0-102.0% (ACS specification)
99%
grade:
ACS reagent
form:
solid

$75.30


In StockDetails


Request a Bulk Order

grade

ACS reagent

Quality Level

vapor density

6 (vs air)

Assay

98.0-102.0% (ACS specification)
99%

form

solid

pH

5.0-8.2 (25 °C, 5%)

mp

89 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.005%
Ca: ≤0.01%
Fe: ≤5 ppm
K: ≤0.005%
Mn: ≤5 ppm
NH4+: ≤0.003%
Na: ≤0.005%
Sr: ≤0.005%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Mg.2NO3.6H2O/c;2*2-1(3)4;;;;;;/h;;;6*1H2/q+2;2*-1;;;;;;

InChI key

MFUVDXOKPBAHMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium nitrate hexahydrate is a hydrated magnesium salt. A mixture of magnesium nitrate hexahydrate and magnesium chloride hexahydrate has been reported as a potential phase change material (PCM) for the storage of latent thermal energy.

Application

Magnesium nitrate hexahydrate can be used:
  • In the synthesis of magnesium oxide (MgO) nanopowder by thermal plasma method.
  • As a starting material in the synthesis of magnesium nitrate – alumina composite [(1-x) Mg (NO3)2 – xAl2O3] via sol-gel method.
  • As a starting material for the synthesis of Mg-Al spinel precursors, required for the preparation of magnesium aluminate spinel (MgAl2O4) powder.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vishwanath Hiremath et al.
Journal of environmental sciences (China), 76, 80-88 (2018-12-12)
The development of carbon dioxide (CO2) sorbents that can operate at elevated temperatures is significant for the advancement of pre-combustion capture technologies. Recently, promoter-based systems composed of alkali/alkaline earth metal nitrates and/or carbonates have been considered as next-generation solid sorbents
Vishwanath Hiremath et al.
ChemSusChem, 12(12), 2810-2818 (2019-04-10)
Although MgO-Al2 O3 is well known as having a spinel structure, the inversion of which occurs by exchange of the trivalent (Al3+ ) and divalent (Mg2+ ) cations, little analytical study of the degree of inversion has been carried out.
Performance of heat charge/discharge of magnesium nitrate hexahydrate and magnesium chloride hexahydrate mixture to a single vertical tube for a latent heat storage system.
Nagano K, et al.
Applied Thermal Engineering, 24(2), 209-220 (2004)
Cell dimensions and space group of magnesium nitrate hexahydrate.
Mozzi RL and Bekebrede WR.
Acta Crystallographica, 14(12), 1296-1297 (1961)
A wet-chemical process yielding reactive magnesium aluminate spinel (MgAl2O4) powder.
Li JG, et al.
Ceramics International, 27(4), 481-489 (2001)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service