Skip to Content
Merck
All Photos(1)

Key Documents

200093

Sigma-Aldrich

Mercury(II) sulfate

ACS reagent, ≥98.0%

Synonym(s):

Mercuric sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgSO4
CAS Number:
Molecular Weight:
296.65
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

ACS reagent

Assay

≥98.0%

reaction suitability

reagent type: catalyst
core: mercury

reduction residue

≤0.02%

anion traces

chloride (Cl-): ≤0.003%
nitrate (NO3-): ≤0.005%

cation traces

Fe: ≤0.005%
Hg+: ≤0.15%

SMILES string

[Hg++].[O-]S([O-])(=O)=O

InChI

1S/Hg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

DOBUSJIVSSJEDA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal analysis of mercury (I) sulfate and mercury (II) sulfate.
Tariq SA and Hill JO.
J. Therm. Anal., 21(2), 277-281 (1981)
Catalysis by mercuric ion of reactions of glycals with water.
Gonzalez F, et al.
Carbohydrate Research, 42(2), 267-274 (1975)
Direct oxidation of methane to methanol by mercuric sulfate catalyst.
Gang X, et al.
J. Catal., 196(2), 287-292 (2000)
Necati Kayaalp et al.
Environmental science and pollution research international, 17(9), 1547-1552 (2010-05-25)
Chemical oxygen demand (COD) is used as a discharge standard parameter in wastewater treatment plant design, environmental modelling and many other applications. Chloride interference is an important problem of COD measurement for wastewaters containing low organic matter and high chloride
L A Ivanova et al.
Gigiena i sanitariia, (11)(11), 66-67 (1991-11-01)
In experiments with two tissue culture lines it was shown, that cytotoxicity of mercury (1) derivatives (measured by CL50) is lesser two times, than it was for mercury (2) derivates. No significant differences in cytotoxicity were noted by the cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service