Skip to Content
Merck
All Photos(1)

Documents

15677

Sigma-Aldrich

Heptane

puriss., ≥99% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
Beilstein:
1730763
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

vapor density

3.5 (vs air)

Quality Level

vapor pressure

40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

grade

puriss.

Assay

≥99% (GC)

form

liquid

autoignition temp.

433 °F

expl. lim.

7 %

impurities

≤0.001% free acid (as CH3COOH)
≤0.0015% non-volatile matter
≤0.02% water (Karl Fischer)

refractive index

n20/D 1.387 (lit.)

bp

98 °C (lit.)

mp

−91 °C (lit.)

density

0.684 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Monadjemi et al.
Environmental science & technology, 45(22), 9582-9589 (2011-09-29)
Photodegradation is seldom considered at the surface of vegetation after crop spraying. Chlorothalonil, a broad-spectrum foliar fungicide with a very widespread use worldwide, was considered. To represent the waxy upper layer of leaves, tests were performed within thin paraffin wax
Thushara Karunasekara et al.
Journal of chromatography. A, 1218(6), 809-816 (2011-01-07)
Partition coefficients for varied compounds were determined for the organic solvent-propylene carbonate biphasic partition system where the organic solvent is n-heptane, isopentyl ether or 1-octanol. These partition coefficient databases are analyzed using the solvation parameter model facilitating a quantitative comparison
Gábor Csordás et al.
PloS one, 9(6), e98191-e98191 (2014-06-04)
In recent years, Drosophila melanogaster has become an attractive model organism in which to study the structure and development of the cellular immune components. The emergence of immunological markers greatly accelerated the identification of the immune cells (hemocytes), while the
Bing Chen et al.
Journal of hazardous materials, 188(1-3), 369-374 (2011-02-26)
The influence of the initial fuel temperature on the burning behavior of n-heptane pool fire was experimentally studied at the State Key Laboratory of Fire Science (SKLFS) large test hall. Circular pool fires with diameters of 100mm, 141 mm, and
Arun K Kota et al.
Advanced materials (Deerfield Beach, Fla.), 24(43), 5838-5843 (2012-08-30)
Hierarchically structured, superoleophobic surfaces are demonstrated that display one of the lowest contact angle hysteresis values ever reported - even with extremely low-surface-tension liquids such as n-heptane. Consequently, these surfaces allow, for the first time, even ≈2 μL n-heptane droplets

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service