Skip to Content
Merck
Heptane biotech. grade, ≥99%

Heptane

Heptane biotech. grade, ≥99%
Synonym(s):
Heptane
Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique