Skip to Content
Merck
All Photos(1)

Key Documents

12619

Sigma-Aldrich

Potassium disulfite

puriss., meets analytical specification of Ph.Eur., BP, NF, FCC, E224, 95.0-101.0% (iodometric), powder, 51.8-57.6% sulfur dioxide

Synonym(s):

Potassium metabisulfite, Potassium pyrosulfite

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$113.00

$113.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
$113.00

About This Item

Linear Formula:
K2S2O5
CAS Number:
Molecular Weight:
222.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
powder

$113.00


Available to ship on14 April 2025Details


Request a Bulk Order

grade

puriss.

Assay

95.0-101.0% (iodometric)

form

powder

quality

meets analytical specification of Ph.Eur., BP, NF, FCC, E224

impurities

organic volatile impurities, complies (GC)
residual solvents, in accordance
≤0.001% heavy metals (as Pb)
≤0.08% thiosulfate (S2O3)
51.8-57.6% sulfur dioxide

pH

3.5-4.5 (20 °C, 5%)

anion traces

chloride (Cl-): ≤100 mg/kg

cation traces

As: ≤2 mg/kg
Cu: ≤20 mg/kg
Fe: ≤10 mg/kg
Hg: ≤1 mg/kg
Pb: ≤2 mg/kg
Se: ≤5 mg/kg
Zn: ≤20 mg/kg

suitability

in accordance for appearance of solution

SMILES string

[K+].[K+].[O-]S(=O)S([O-])(=O)=O

InChI

1S/2K.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

RWPGFSMJFRPDDP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bindu Nair et al.
International journal of toxicology, 22 Suppl 2, 63-88 (2003-10-14)
Sodium Sulfite, Ammonium Sulfite, Sodium Bisulfite, Potassium Bisulfite, Ammonium Bisulfite, Sodium Metabisulfite, and Potassium Metabisulfite are inorganic salts that function as reducing agents in cosmetic formulations. All except Sodium Metabisulfite also function as hair-waving/straightening agents. In addition, Sodium Sulfite, Potassium
Hisayori Shigechi et al.
Applied and environmental microbiology, 70(8), 5037-5040 (2004-08-06)
Direct and efficient production of ethanol by fermentation from raw corn starch was achieved by using the yeast Saccharomyces cerevisiae codisplaying Rhizopus oryzae glucoamylase and Streptococcus bovis alpha-amylase by using the C-terminal-half region of alpha-agglutinin and the flocculation functional domain
Gábor Vlád et al.
The Journal of organic chemistry, 67(18), 6550-6552 (2002-08-31)
A high-yield synthesis was developed for the preparation of 2,2'-bipyrimidine (1) using the Ullmann coupling of 2-iodopyrimidine. The new procedure was also used for the preparation of 4,4',6,6'-tetramethyl-2,2'-bipyrimidine (2) and 5,5'-dibromo-2,2'-bipyrimidine (3).
S Raisuddin et al.
Food additives and contaminants, 8(6), 707-712 (1991-11-01)
The occurrence of aflatoxins in market betel nut samples was studied. It was observed that several betel nut samples were infested with aflatoxin-producing fungus, Aspergillus flavus. Out of 32 samples collected from various places, 12 were positive for aflatoxin. Aflatoxin
D A Mathison et al.
Chest, 87(1 Suppl), 50S-54S (1985-01-01)
Several types of reactions to drugs and chemicals may precipitate or perpetuate asthmatic relapse. This review focuses on reactions to aspirin and sulfites. Approximately 40 percent of patients with rhinosinusitis, nasal polyps, and asthma and 5 to 10 percent of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service