Skip to Content
Merck
All Photos(1)

Key Documents

Y0000461

Torasemide anhydrous

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Torsemide, Demadex, N-[[(1-Methylethyl)amino]carbonyl]-4-[(3-methylphenyl)amino]-3-pyridinesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

30 MG
$254.00

$254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
30 MG
$254.00

About This Item

Empirical Formula (Hill Notation):
C16H20N4O3S
CAS Number:
Molecular Weight:
348.42
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$254.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

torasemide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)NC(=O)NS(=O)(=O)c1cnccc1Nc2cccc(C)c2

InChI

1S/C16H20N4O3S/c1-11(2)18-16(21)20-24(22,23)15-10-17-8-7-14(15)19-13-6-4-5-12(3)9-13/h4-11H,1-3H3,(H,17,19)(H2,18,20,21)

InChI key

NGBFQHCMQULJNZ-UHFFFAOYSA-N

Gene Information

human ... SLC12A1(6557)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Torasemide anhydrous EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Torsemide is a loop diuretic and inhibitor of the Na+/K+/2Cl- carrier system.
Torsemide is a loop diuretic of the pyridine-sulfonylurea class with antialdosteronergic properties and inhibitor of the Na+/K+/2Cl- carrier system.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agents with antialdosterone properties should be the preferred diuretics for reducing hypertension related atrial fibrillation.
Oscar M P Jolobe
European journal of internal medicine, 21(1), 55-55 (2010-02-04)
[Treatment of nephritic edema by torasemide in children].
Qi Li et al.
Zhongguo dang dai er ke za zhi = Chinese journal of contemporary pediatrics, 13(6), 520-521 (2011-06-16)
Clinical therapeutics, 33(9), 1204-1213 (2011-09-13)
The pharmacologic modification of the synthesis and deposition of fibrillar collagen in the myocardium may have effects on the cardiac function, clinical status, and prognosis of patients with heart failure (HF). Serum procollagen type I carboxyterminal peptide (PICP) is a
[Etiologies of nephrotic syndrome].
S Heeringa et al.
Praxis, 100(9), 505-514 (2011-04-29)
José C Rodríguez-Pérez et al.
Clinical and experimental hypertension (New York, N.Y. : 1993), 33(8), 506-510 (2011-10-01)
We evaluated the anti-hypertensive and anti-albuminuric effect of the angiotensin receptor blocker telmisartan alone and in combination with torasemide and amlodipine. Patients were hypertensive, both diabetics and non-diabetics with persistent microalbuminuria. Our primary endpoint was a change in microalbuminuria levels

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service