Skip to Content
Merck
All Photos(1)

Key Documents

Y0000207

Perindopril for stereochemical purity

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Perindopril erbumine, (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylic acid tert-butylamine salt, Perindopril tert-butylamine, S-9490

Sign Into View Organizational & Contract Pricing

Select a Size

40 MG
$221.00

$221.00


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
40 MG
$221.00

About This Item

Empirical Formula (Hill Notation):
C19H32N2O5 · C4H11N
CAS Number:
Molecular Weight:
441.60
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$221.00


Available to ship on15 April 2025Details


Request a Bulk Order

grade

pharmaceutical primary standard

API family

perindopril

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(C)N.CCC[C@H](N[C@@H](C)C(=O)N1[C@H]2CCCC[C@H]2C[C@H]1C(O)=O)C(=O)OCC

InChI

1S/C19H32N2O5.C4H11N/c1-4-8-14(19(25)26-5-2)20-12(3)17(22)21-15-10-7-6-9-13(15)11-16(21)18(23)24;1-4(2,3)5/h12-16,20H,4-11H2,1-3H3,(H,23,24);5H2,1-3H3/t12-,13-,14-,15-,16-;/m0./s1

InChI key

IYNMDWMQHSMDDE-MHXJNQAMSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Perindopril for stereochemical purity EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Perindopril erbumine is an angiotensin converting enzyme (ACE) inhibitor; antihypertensive.
Perindopril erbumine is an angiotensin converting enzyme (ACE) inhibitor; antihypertensive; becomes hydrolyzed in vivo to the active diacid metabolite; unlike the other ACE inhibitors, inhibits tumor growth in hepatocellular carcinoma cells due to suppression of VEGF levels; also suppresses angiotensin II production in vitro. Long-term therapy with this agent has a beneficial effect on the cerebral circulation by improving cerebral perfusion reserve in patients with previous minor stroke.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arthur J A Leloup et al.
Hypertension (Dallas, Tex. : 1979), 64(1), 195-200 (2014-04-23)
Arterial stiffening is the root cause of a range of cardiovascular complications, including myocardial infarction, left ventricular hypertrophy, stroke, renal failure, dementia, and death, and a hallmark of the aging process. The most important in vivo parameter of arterial stiffness
Fei Xiong et al.
The Journal of surgical research, 189(1), 166-173 (2014-03-08)
The purpose of the present study was to evaluate the effect of a new angiotensin converting enzyme inhibitor perindopril on the formation of experimental abdominal aortic aneurysms (AAAs) in a rat model induced by intraluminal elastase infusion and extraluminal calcium
Sophia Zoungas et al.
The New England journal of medicine, 371(15), 1392-1406 (2014-09-23)
In the Action in Diabetes and Vascular Disease: Preterax and Diamicron Modified Release Controlled Evaluation (ADVANCE) factorial trial, the combination of perindopril and indapamide reduced mortality among patients with type 2 diabetes, but intensive glucose control, targeting a glycated hemoglobin
Haruka Nishimuta et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(9), 1522-1531 (2014-07-06)
Hydrolysis plays an important role in metabolic activation of prodrugs. In the current study, species and in vitro system differences in hepatic and extrahepatic hydrolysis were investigated for 11 prodrugs. Ten prodrugs in the data set are predominantly hydrolyzed by
Joep van der Leeuw et al.
Hypertension (Dallas, Tex. : 1979), 65(1), 115-121 (2014-10-15)
Blood pressure-lowering treatment reduces cardiovascular risk in patients with diabetes mellitus, but the effect varies between individuals. We sought to identify which patients benefit most from such treatment in a large clinical trial in type 2 diabetes mellitus. In Action

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service