Skip to Content
Merck
All Photos(1)

Key Documents

W245735

Sigma-Aldrich

Ethyl pyruvate

≥97%, FG

Synonym(s):

Ethyl 2-oxopropanoate, Ethyl 2-oxopropionate, Ethyl acetyl formate, Ethyl pyroracemate, Pyruvic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

W245735-SAMPLE-K
$97.10
250 G
$143.00
1 KG
$279.00
10 KG
$1,760.00
25 KG
$4,120.00

$97.10


Estimated to ship on31 March 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W245735-SAMPLE-K
$97.10
250 G
$143.00
1 KG
$279.00
10 KG
$1,760.00
25 KG
$4,120.00

About This Item

Linear Formula:
CH3COCOOC2H5
CAS Number:
Molecular Weight:
116.12
FEMA Number:
2457
Beilstein:
1071466
EC Number:
Council of Europe no.:
430
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.442
NACRES:
NA.21
Organoleptic:
fruity; ethereal; rum; sweet
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

$97.10


Estimated to ship on31 March 2025


Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.404 (lit.)

bp

144 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fruity; ethereal; rum; sweet

SMILES string

CCOC(=O)C(C)=O

InChI

1S/C5H8O3/c1-3-8-5(7)4(2)6/h3H2,1-2H3

InChI key

XXRCUYVCPSWGCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • High mobility group box 1 in the central nervous system: regeneration hidden beneath inflammation.: This study explores the role of high mobility group box 1 (HMGB1) in the central nervous system, focusing on its regenerative capabilities despite its inflammatory properties. Ethyl pyruvate was identified as a potential therapeutic agent in mitigating inflammation and promoting neural regeneration (Kim et al., 2025).

  • Immunomodulation of Ethyl pyruvate on gestational diabetes mellitus in Mice; The impact on Th17/Treg balance.: This study investigates the effects of ethyl pyruvate on gestational diabetes mellitus (GDM) in mice, focusing on its immunomodulatory effects and its impact on the Th17/Treg balance, highlighting its potential benefits in managing GDM (Zhao et al., 2024).

  • HMGB1/RAGE axis in tumor development: unraveling its significance.: The review delves into the HMGB1/RAGE axis in tumor development, examining its significance and the potential therapeutic role of ethyl pyruvate in targeting this pathway to inhibit tumor growth and progression (Fan et al., 2024).

  • Ethyl pyruvate ameliorates acute respiratory distress syndrome in mice.: This article highlights the therapeutic potential of ethyl pyruvate in ameliorating acute respiratory distress syndrome (ARDS) in a mouse model, showcasing its anti-inflammatory and protective effects (Micanovic et al., 2024).

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service