Skip to Content
Merck
All Photos(1)

Key Documents

8.06617

Sigma-Aldrich

Ethyl pyruvate

for synthesis

Synonym(s):

Ethyl pyruvate, Pyruvic acid ethyl ester, Ethyl 2-oxopropanoate, Ethyl 2-oxopropionate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$94.70
500 ML
$335.00

$94.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$94.70
500 ML
$335.00

About This Item

Empirical Formula (Hill Notation):
C5H8O3
CAS Number:
Molecular Weight:
116.12
MDL number:
UNSPSC Code:
12352108
EC Index Number:
210-511-2
NACRES:
NA.22

$94.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

liquid

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

bp

144-146 °C/1013 hPa

mp

-58 °C

transition temp

flash point 52 °C

density

1.05 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

O(CC)C(=O)C(=O)C

InChI

1S/C5H8O3/c1-3-8-5(7)4(2)6/h3H2,1-2H3

InChI key

XXRCUYVCPSWGCC-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.044 - 1.049
Identity (IR): passes test

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service