The enantiomeric excess (ee) is not determined. The optical rotation specification (neat) is -5.5 to -3.5°.
W230920
(S)-(−)-β-Citronellol
99%, FG
Synonym(s):
β-Rhodinol, (S)-3,7-Dimethyl-6-octen-1-ol
Select a Size
Select a Size
About This Item
Halal
Kosher
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Halal
Kosher
reg. compliance
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
vapor pressure
~0.02 mmHg ( 25 °C)
Assay
99%
refractive index
n20/D 1.456 (lit.)
bp
225-226 °C (lit.)
density
0.856 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
floral; waxy; rose
SMILES string
C[C@H](CCO)CC\C=C(\C)C
InChI
1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3/t10-/m0/s1
InChI key
QMVPMAAFGQKVCJ-JTQLQIEISA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
213.8 °F - DIN 51758
Flash Point(C)
101 °C - DIN 51758
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
what is the enantiomeric excess of the W930920 (S)-citronellol?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service