Skip to Content
Merck
All Photos(1)

Key Documents

533904

Sigma-Aldrich

1-Bromo-3,7-dimethyloctane

96%

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$242.00
25 ML
$787.00

$242.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$242.00
25 ML
$787.00

About This Item

Linear Formula:
CH2BrCH2CH(CH3)CH2CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
221.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$242.00


Check Cart for Availability

Request a Bulk Order

Assay

96%

refractive index

n20/D 1.455 (lit.)

bp

135-140 °C/10 mmHg (lit.)
216-217 °C (lit.)

density

1.066 g/mL at 20 °C

functional group

alkyl halide
bromo

SMILES string

CC(C)CCCC(C)CCBr

InChI

1S/C10H21Br/c1-9(2)5-4-6-10(3)7-8-11/h9-10H,4-8H2,1-3H3

InChI key

VGSUDZKDSKCYJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.0 °F - closed cup

Flash Point(C)

98.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinjun Shao et al.
Organic & biomolecular chemistry, 10(35), 7045-7052 (2012-08-01)
A series of linear and star-shaped pyrazine-containing acene molecules 1a-b, 2a-b and 3 substituted by dicarboximide groups are synthesized via condensation reactions between o-diamine and dione. High electron affinity (up to 4.01 eV) is achieved due to the introduction of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service