Skip to Content
Merck
All Photos(1)

Key Documents

TSS1000

Supelco

Total Suspended Solids 1000 mg/L Calibration Standard

certified reference material

Synonym(s):

TSS Reference Solution (1000 mg/L)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$113.00
500 ML
$150.00

$113.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$113.00
500 ML
$150.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$113.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC TSS1000-100

concentration

in H2O

application(s)

environmental

format

single component solution

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Total Suspended Solids 1000 mg/L Calibration Standard for R&D and Analytical use only.

Features and Benefits

  • Packed in High Density Polyethylene (HDPE) bottle
  • A certificate of analysis (CoA) is included with the product
  • Traceable to recognized national standards, and units of measurement recognized at the National Institute of Standards and Technology (NIST) and International System of units (SI)
  • Gravimetrically performed dilutions
  • Balance used to prepare the standard is calibrated to ISO 17025 requirements
  • Stable solution with long minimum shelf life

Other Notes

Values of analytes vary lot to lot.

related product

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service