Skip to Content
Merck
All Photos(1)

Key Documents

S5753

Supelco

β-Sitosterol

analytical standard, from soybean, ≥40%

Synonym(s):

α-Dihydrofucosterol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol, 5-Stigmasten-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O
CAS Number:
Molecular Weight:
414.71
Beilstein:
1916165
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

biological source

soybean

grade

analytical standard

concentration

≥40%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

136-140 °C (lit.)

application(s)

food and beverages

format

neat

functional group

hydroxyl

shipped in

ambient

storage temp.

room temp

SMILES string

O[C@H](C1)CC[C@@]2(C)C1=CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CC[C@H](C(C)C)CC

InChI

1S/C29H50O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-21,23-27,30H,7-9,11-18H2,1-6H3/t20-,21-,23+,24+,25-,26+,27+,28+,29-/m1/s1

InChI key

KZJWDPNRJALLNS-VJSFXXLFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Quality

Practical
Also contains campesterol and dihydrobrassicasterol as identified by 13C-NMR.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koizumi, N., et al.
Chemical & Pharmaceutical Bulletin, 27, 38-38 (1979)
Wright, J.L.C., et al.
Canadian Journal of Chemistry, 56, 1898-1898 (1978)
Günther Silbernagel et al.
Current opinion in lipidology, 24(1), 12-17 (2012-11-21)
Plant sterols as ingredients to functional foods are recommended for lowering LDL cholesterol. However, there is an ongoing discussion whether the use of plant sterols is safe. Genetic analyses showed that common variants of the ATP binding cassette transporter G8
Ashutosh Singh
Food chemistry, 137(1-4), 62-67 (2012-12-04)
The antioxidative effect of sitosterol at 1, 2 and 5% levels, in triolein, refined canola, high oleic sunflower and flaxseed oils, continuously heated for a period of up to 72 h at frying temperature of 180 °C, was studied. High
Daniele Silvestro et al.
PloS one, 8(2), e56429-e56429 (2013-02-15)
Sterols are crucial lipid components that regulate membrane permeability and fluidity and are the precursors of bioactive steroids. The plant sterols exist as three major forms, free sterols, steryl glycosides and steryl esters. The storage of steryl esters in lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service