Skip to Content
Merck
All Photos(1)

Key Documents

567152

Sigma-Aldrich

β-Sitosterol

≥75% (β-sitosterol, GC), solid, anticholesteremic steroid, Calbiochem®

Synonym(s):

β-Sitosterol, α-Dihydrofucosterol, 22,23-Dihydrostigmasterol, α-Phytosterol, SITO

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$76.10
25 G
$346.00

$76.10


Available to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$76.10
25 G
$346.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$76.10


Available to ship on01 April 2025Details


Request a Bulk Order

Product Name

β-Sitosterol, Common plant steroid that possesses anticholesteremic properties.

Quality Level

description

Merck USA index - 14, 8556

Assay

≥75% (β-sitosterol, GC)
≥95% sterol basis

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤2.5% (GC)

color

white

solubility

chloroform: 5 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

O[C@H]1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@H](CC[C@H](C(C)C)CC)C)(CC3)C)CC=C2C1)C

InChI

1S/C29H50O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-21,23-27,30H,7-9,11-18H2,1-6H3/t20-,21-,23+,24+,25-,26+,27+,28+,29-/m1/s1

InChI key

KZJWDPNRJALLNS-VJSFXXLFSA-N

General description

Common plant steroid that possesses anticholesteremic properties. Has recently been shown to induce G2/M arrest, endoreduplication, and apoptosis through the Bcl-2 and PI3K/Akt signaling pathways.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Anticholesteremic properties
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Other Notes

Moon, D., et al. 2008. Cancer Lett.264, 181.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service