Skip to Content
Merck
All Photos(4)

Key Documents

S2378

Sigma-Aldrich

Sodium succinate dibasic hexahydrate

ReagentPlus®, ≥99%

Synonym(s):

Butanedioic acid disodium salt, Succinic acid disodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$67.60
500 G
$198.00
1 KG
$266.00

$67.60


Available to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$67.60
500 G
$198.00
1 KG
$266.00

About This Item

Linear Formula:
NaOOCCH2CH2COONa · 6H2O
CAS Number:
Molecular Weight:
270.14
Beilstein:
3921044
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
solubility:
H2O: 100 mg/mL, clear, colorless

$67.60


Available to ship on04 April 2025Details


Request a Bulk Order

grade

reagent

Quality Level

product line

ReagentPlus®

Assay

≥99%

solubility

H2O: 100 mg/mL, clear, colorless

SMILES string

O.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O

InChI

1S/C4H6O4.2Na.6H2O/c5-3(6)1-2-4(7)8;;;;;;;;/h1-2H2,(H,5,6)(H,7,8);;;6*1H2/q;2*+1;;;;;;/p-2

InChI key

ZBTUYCUNQBRXOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Sodium succinate dibasic hexahydrate can be used as an organic additive in the preparation of well-defined hollow nano-baskets morphology of ZnO nanoparticles (ZnO-HNBs) via a low-temperature solution method using zinc nitrate hexahydrate and hexamethylenetetramine. ZnO-HNBs are further applied as photocatalysts for dye degradation.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Seop Ahn et al.
International journal of molecular sciences, 19(10) (2018-10-03)
Myosin heavy chain (MyHC) isoforms consist of Myh7, Myh2, Myh1, and Myh4, which are expressed in skeletal muscle tissues during postnatal development. These genes influence the contraction⁻relaxation activity in skeletal muscles and are involved in determining muscle composition such as
Oswaldo A Lozoya et al.
Cell reports, 32(11), 108131-108131 (2020-09-17)
Mitochondrial-driven alterations of the epigenome have been reported, but whether they are relevant at the organismal level remains unknown. The viable yellow agouti mouse (Avy) is a powerful epigenetic biosensor model that reports on the DNA methylation status of the
Jean-Philippe Leduc-Gaudet et al.
The Journal of physiology, 597(7), 1975-1991 (2019-01-08)
Recent evidence suggests that impaired mitophagy, a process in charge of removing damaged/dysfunctional mitochondria and in part regulated by Parkin, could contribute to the ageing-related loss of muscle mass and function. In the present study, we show that Parkin overexpression
Thamir Alandijany et al.
Journal of neurovirology, 19(6), 537-549 (2013-11-28)
Infection with the challenge virus standard-11 (CVS) strain of fixed rabies virus induces neuronal process degeneration in adult mice after hindlimb footpad inoculation. CVS-induced axonal swellings of primary rodent dorsal root ganglion neurons are associated with 4-hydroxy-2-nonenal protein adduct staining
Marta Murgia et al.
Cell reports, 29(12), 3825-3834 (2019-12-19)
The mosaic distribution of cytochrome c oxidase+ (COX+) and COX- muscle fibers in mitochondrial disorders allows the sampling of fibers with compensated and decompensated mitochondrial function from the same individual. We apply laser capture microdissection to excise individual COX+ and

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service