Skip to Content
Merck
All Photos(4)

Documents

224731

Sigma-Aldrich

Succinic acid disodium salt

99%

Synonym(s):

Sodium succinate dibasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCH2CH2COONa
CAS Number:
Molecular Weight:
162.05
Beilstein:
3917246
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

SMILES string

[Na+].[Na+].[O-]C(=O)CCC([O-])=O

InChI

1S/C4H6O4.2Na/c5-3(6)1-2-4(7)8;;/h1-2H2,(H,5,6)(H,7,8);;/q;2*+1/p-2

InChI key

ZDQYSKICYIVCPN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Succinic acid disodium salt is used to prepare buffer solution for the quantitative analysis of auxins.
It can be also used:        
  • To study the effect of liquid phase on the basic properties of α-tricalcuim phosphate (α-TCP) based cement, BIOPEX.        
  • In the synthesis of cadmium sulfide (CdS) nanoparticles using thiourea as a source for sulfide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterisation and photocatalytic activity of cadmium sulphide nanoparticles
Kakodkar SB
Chemical Science Transactions, 5, 75-78 (2016)
Abubakr Mossa et al.
Neurourology and urodynamics, 40(1), 120-130 (2020-10-25)
Succinate activates the receptor GPR91 identified in the bladder. The present study aims to unravel the mechanisms of bladder relaxation by succinate and how the receptor is involved in structural and functional changes of the bladder. Physiological recordings of bladder
Effects of liquid phase on basic properties of ?-tricalcium phosphate-based apatite cement
O Makoto, et al.
Dental Materials Journal, 27(5), 672-677 (2008)
Mohammed Z Allouh et al.
Histology and histopathology, 32(11), 1151-1159 (2017-01-20)
Sustanon is a well-known anabolic drug that is used to treat hypogonadism and restore muscle mass and bone density. As research to date has been limited to its effects in glycolytic fibers, this study aimed to investigate the dose-related effects
Makoto Oda et al.
Dental materials journal, 27(5), 672-677 (2008-11-01)
Effects of liquid phase on the basic properties of alpha-tricalcuim phosphate (alpha-TCP)-based cement, BIOPEX, were investigated by employing three liquid phases: distilled water, neutral sodium hydrogen phosphate solution, and succinic acid disodium salt solution containing sodium salt of chondroitin sulfate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service