Skip to Content
Merck
All Photos(1)

Key Documents

RTC000087

Supelco

Ethyl Acetate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ethyl acetate, EtOAc

Sign Into View Organizational & Contract Pricing

Select a Size

20 ML
$224.00

$224.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
20 ML
$224.00

About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
MDL number:
UNSPSC Code:
41116107

$224.00


Available to ship on22 April 2025Details


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1265402

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

packaging

pkg of 20 mL

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

application(s)

pharmaceutical small molecule

format

neat

storage temp.

2-30°C

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Analysis Note

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB9766 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

26.6 °F

Flash Point(C)

-3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service