Skip to Content
Merck
All Photos(2)

Key Documents

1.10972

Supelco

Ethyl acetate

for gas chromatography ECD and FID SupraSolv®

Synonym(s):

Ethyl ethanoate, Acetic ester, Ethyl ester, EtOAc

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$74.70
2.5 L
$156.00
4 L
$260.00

$74.70


Estimated to ship on27 March 2025


Request a Bulk Order

Select a Size

Change View
1 L
$74.70
2.5 L
$156.00
4 L
$260.00

About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
MDL number:
UNSPSC Code:
12191502
EC Index Number:
205-500-4
NACRES:
NA.03
Assay:
≥99.8% (GC)
technique(s):
gas chromatography (GC): suitable
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)

$74.70


Estimated to ship on27 March 2025


Request a Bulk Order

vapor density

3 (20 °C, vs air)

Quality Level

vapor pressure

73 mmHg ( 20 °C)

product line

SupraSolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

801 °F

potency

5620 mg/kg LD50, oral (Rat)
>18000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.2-11.5 %, 38 °F

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

transition temp

flash point -4 °C

solubility

85.3 g/L

density

0.902 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SupraSolv ECD and FID are specially developed for gas chromatography in combination with the ECD (Electron Capture Detector) and FID (Flame Ionization Detector). SupraSolv solvents offer the largest specified retention time range, a clear baseline, and a minimal signal-to-noise ratio, leading to reliable and reproducible analysis results.

Application

Ethyl acetate can be used as a:
  • Extraction solvent for the pharmaceutical metabolites by sonication in an ultrasonic bath.[1]
  • Rinsing solvent for glassware.[2]

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of phthalates in milk and milk products by liquid chromatography/tandem mass spectrometry
Sorensen LK
Rapid Communications in Mass Spectrometry, 20(7), 1135-1114 (2006)
Photodegradation of Carbamazepine, Ibuprofen, Ketoprofen and 17?-Ethinylestradiol in Fresh and Seawater
Matamoros V, et al.
Water, Air, and Soil Pollution, 196, 161-116 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service