Skip to Content
Merck
All Photos(2)

Documents

RDD036

Sigma-Aldrich

BICINE

Redi-Dri, ≥99%, free-flowing

Synonym(s):

N,N-Bis(2-hydroxyethyl)glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO4
CAS Number:
Molecular Weight:
163.17
Beilstein:
1769362
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.21

product line

Redi-Dri

Quality Level

Assay

≥99%

quality

free-flowing

useful pH range

7.6-9.0
7.6-9.0

pKa (25 °C)

8.3

SMILES string

OCCN(CCO)CC(O)=O

InChI

1S/C6H13NO4/c8-3-1-7(2-4-9)5-6(10)11/h8-9H,1-5H2,(H,10,11)

InChI key

FSVCELGFZIQNCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bicine is an inorganic compound used to make Bicine buffer, a "Good buffer" that is commonly used in molecular biology and biochemistry. Bicine is a zwitterionic substituted glycine that is stable and soluble in water. It is has a pKa of 8.3, ideal for biological sytems with a pH range of 7.6 - 9.0.

Application

Redi-Dri is an innovative packaging product line that helps eliminate clumping material by adding extra packaging process steps without any chemical additives, anti-clumping agents, or hydrophobic compounds. Redi Dri offers a more convenient and safer way to handle the products.
Bicine is typically used for protein analysis:
  • for SDS-PAGE or Western blot assay, for column wash buffer, to pH solutions, to make digestion buffer for GFP & FRET Microscopy
  • labeling buffer to prepare antibody solution for labeing for fluorecent assays
Recommended buffer for low temperature biochemical work. Used in the preparation of stable substrate solution for serum guanase determination. Spacer in plasma protein fractionation with Sephadex® by isotachophoresis.

Features and Benefits

  • Redi-Dri design controls moisture for hygroscopic chemicals that commonly clump
  • Anhydrous powder remains free-flowing and easy to use
  • Eliminates clumps, for safer routine use in lab
  • Maintains product moisture, quality, and attributes

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC
Sephadex is a registered trademark of Cytiva

comparable product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imaging protein interactions by FRET microscopy: labeling proteins with fluorescent dyes
Verveer PJ et al.
CSH Protocols, 2006, pdb-pdb (2006)
Probing Protein Interactions Using GFP and FRET * Stage 1: Labeling Proteins with Fluorescent Dyes
Joseph Sambrook, David W Russell
CSH Protocols, 2006, pdb-pdb (2006)

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service