Skip to Content
Merck
All Photos(3)

Key Documents

PPB019

Sigma-Aldrich

Bis-Tris Bicine EDTA buffer

pH 7.2, pHast Pack, powder

Synonym(s):

1X Bis-Tris transfer buffer, Bis-Tris

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
$187.00

$187.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 PKG
$187.00

About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

$187.00


Check Cart for Availability

Request a Bulk Order

product line

pHast Pack

Quality Level

form

powder

storage condition

(Tightly closed. Dry)

pH

7.2

cation traces

Pb: ≤5 ppm

foreign activity

DNAse, free
NICKase, free
Protease, free
RNAse, free

storage temp.

room temp

General description

Bis-Tris Bicine EDTA buffer or Bis-Tris buffer (pH 7.2) contains Bis-Tris and Bicine, zwitterionic buffer components that form strong bonds with common metals. In combination with ethylenediaminetetraacetic acid (EDTA), a metal chelator, that binds divalent cations. This buffer is commonly used as a transfer buffer in western blot to transfer proteins from the gel to the membrane.

Application

Bis-Tris buffer may be used in western blot as a transfer buffer.

Features and Benefits

  • Ready-to-use buffer
  • No measuring and pH adjustment needed
  • Certified quality for reproducibility
  • Minimizes exposure to toxic chemicals
  • Biological tests: free of DNase, RNase, Protease, and Nickase
  • Chemical tests: Iron ≤10 ppm, Lead ≤5 ppm

Packaging

Foil pouches

Reconstitution

Contents of one pouch, when dissolved in 500 mL of ultrapure water, will yield a 1X solution containing 25 mM Bicine, 25 mM Bis-Tris, 1 mM EDTA, pH 7.2 at 25 °C. Contents tested to be DNAse, RNAse, Protease, and Nickase free.

Storage and Stability

Store at room temperature. Product may naturally agglomerate but can be simply broken up within the pouch prior to use.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

pHast Pack is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service