Skip to Content
Merck
All Photos(1)

Key Documents

QC1134

Supelco

Total Phenolics

certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$76.20

$76.20


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
2 ML
$76.20

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$76.20


Estimated to ship on30 May 2025


Request a Bulk Order

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC QCI-032

concentration

in acetone

technique(s)

UV/Vis spectroscopy: suitable

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup - Solvent

Flash Point(C)

-17 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weimin Peng et al.
Genetics, 179(1), 277-289 (2008-05-22)
Histone acetylation levels are regulated through the opposing activities of histone acetyltransferases (HATs) and deacetylases (HDACs). While much is known about gene-specific control of histone acetylation, little is understood about how total or cellular levels of histone acetylation are regulated.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service