Skip to Content
Merck
All Photos(1)

Key Documents

47899

Supelco

EPA 8040A Phenol Calibration Mix

500 μg/mL each component in isopropanol, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$112.00

$112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$112.00

About This Item

UNSPSC Code:
12352200

$112.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Agency

EPA 8040

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

500 μg/mL each component in isopropanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

-10 to -25°C

Application

EPA 8040A phenol calibration mix has been used as an analytical standard in the determination of chlorophenols present in landfill leachates using gas chromatography coupled with mass spectrometry (GC-MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    4-Chloro-3-methylphenol
    2-Chlorophenol
    m-Cresol
    p-Cresol
    2,4-Dichlorophenol
    2,6-Dichlorophenol
    2,4-Dimethylphenol
    2,4-Dinitrophenol
    Dinoseb
    2-Methyl-4,6-dinitrophenol
    o-Cresol
    2-Nitrophenol
    4-Nitrophenol
    Pentachlorophenol
    Phenol
    2,3,4,5-Tetrachlorophenol
    2,3,4,6-Tetrachlorophenol
    2,3,5,6-Tetrachlorophenol
    2,3,4-Trichlorophenol
    2,3,5-Trichlorophenol
    2,3,6-Trichlorophenol
    2,4,5-Trichlorophenol
    2,4,6-Trichlorophenol
    3,4,5-Trichlorophenol
    See All (24)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct determination of chlorophenols in landfill leachates by solid-phase micro-extraction?gas chromatography?mass spectrometry
Ribeiro.A, et al.
Journal of Chromatography A, 975, 267-274 (2002)
Direct determination of chlorophenols in landfill leachates by solid-phase micro-extraction-gas chromatography-mass spectrometry
Ribeiro.A, et al.
Journal of Chromatography A, 975(2), 267-274 (2002)
A Ribeiro et al.
Journal of chromatography. A, 975(2), 267-274 (2002-11-29)
Landfill leachates represent a serious environmental concern with regard to trace priority pollutants introduced into the aquatic environment. From the analytical point of view, they constitute complex matrices because of their high organic matter content and competition with the trace

Questions

1–3 of 3 Questions  
  1. What is Product No. 47889 EPA 8040A Phenol Calibration Mix soluble in?

    1 answer
    1. Product No. 47889 EPA 8040A Phenol Calibration Mix is slightly soluble in water and soluble in alcohol, diethyl ether, and acetone

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. How is Product No. 47889 EPA 8040A Phenol Calibration Mix packaged?

    1 answer
    1. Product No. 47889 comes packaged in a 4ml amber vial with teflon lined solid cap.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service