Skip to Content
Merck
All Photos(1)

Key Documents

PSRVD04

Multi-Parameter Refractive Index & Density Certified Reference Material at 15 °C, 20 °C and 25 °C, Single 30 mL

Nominal RI Value 1.6579 at 20 °C

Sign Into View Organizational & Contract Pricing

Select a Size

30 ML
$690.00

$690.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
30 ML
$690.00

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$690.00


Check Cart for Availability

Request a Bulk Order

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service