Skip to Content
Merck
All Photos(1)

Key Documents

1.08962

Supelco

Refractive index standard kit 1

2,2,4-Trimethyl-pentane/water, traceable to NIST, traceable to PTB, n20/D 1.3915, Certipur®

Synonym(s):

Refraction Calibration Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$576.00

$576.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1 SET
$576.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$576.00


Estimated to ship on25 May 2025


Request a Bulk Order

Agency

traceable to NIST
traceable to PTB

Quality Level

product line

Certipur®

composition

2,2,4-Trimethyl-pentane/water

storage temp.

15-25°C

General description

2,2,4-Trimethyl-pentane/water, traceable to SRM from NIST and PTB n (20°C)=1,3915 Certipur®

Application

Refractive index standard kit 1 for use as a calibration standard for refractometers.

Features and Benefits

  • Five 8ml ampoules of the standard solution and five 8ml ampoules with water are included in each box
  • Traceable to recognized national standards, and to units of measurement recognized at the National Institute Standards and Technology (NIST) and Physical Technical Institute (PTB)
  • Solutions that are stable, ready to use, and have long minimum shelf lives
  • A certificate of analysis (CoA) is included with every standard kit

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service