Skip to Content
Merck
All Photos(1)

Key Documents

PHR2134

4-Isobutylstyrene

Synonym(s):

4-Isobutylstyrene

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$974.00

$974.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$974.00

About This Item

Empirical Formula (Hill Notation):
C12H16
Molecular Weight:
160.26
UNSPSC Code:
41116107
NACRES:
NA.24

$974.00


Check Cart for Availability

Request a Bulk Order

API family

ibuprofen

Quality Level

form

liquid

storage temp.

2-30°C

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Ibuprofen and impurity separation using Ascentis Express 90Å C18 column with excellent peak shape and low quantification limits.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service