Skip to Content
Merck
All Photos(1)

Key Documents

PHR1964

Supelco

Ibuprofen Impurity F

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

3-(4-Isobutylphenyl)propionic acid, 3-[4-(2-METHYLPROPYL)PHENYL] PROPANOIC ACID

Sign Into View Organizational & Contract Pricing

Select a Size

30 MG
$527.00

$527.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
30 MG
$527.00

About This Item

Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
UNSPSC Code:
41116107
NACRES:
NA.24

$527.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000140

API family

ibuprofen

CofA

current certificate can be downloaded

packaging

pkg of 30 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)CCc1ccc(cc1)CC(C)C

InChI

1S/C13H18O2/c1-10(2)9-12-5-3-11(4-6-12)7-8-13(14)15/h3-6,10H,7-9H2,1-2H3,(H,14,15)

InChI key

DYNVRFFVBZVRND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ibuprofen Impurity F is an impurity of the drug, ibuprofen. Ibuprofen is a non-steroidal anti-inflammatory drug (NSAID) that is effective against pain, inflammation and fever. It is deemed as a safe drug on account of its reversible inhibitory effects on cyclooxygenase (COX) enzyme along with its unique pharmacokinetics.[1]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Ibuprofen may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by analytical techniques.[2][3][4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB2590 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Determination of enantiomeric composition of ibuprofen in pharmaceutical formulations by partial least-squares regression of strongly overlapped chromatographic profiles
Grisales JO, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 910, 78-83 (2012)
Two liquid chromatographic methods for the simultaneous determination of ibuprofen and methocarbamol or chlorzoxazone in the presence of their degradation products
Elkady EF and Fouad MA
Journal of Liquid Chromatography and Related Technologies, 35(7), 882-895 (2012)
Ibuprofen: A critical bibliographic review (1999)
Ibuprofen
USP42-NF37: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 34(4), 2232-2232 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service