Skip to Content
Merck
All Photos(1)

Key Documents

PHR1600

Supelco

Topiramate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Topiramate, 2,3:4,5-Bis-O-(1-methylethylidene)-36-D-fructo-pyranose sulfamate, McN 4853, RWJ 17021, Topamax

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$205.00

$205.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$205.00

About This Item

Empirical Formula (Hill Notation):
C12H21NO8S
CAS Number:
Molecular Weight:
339.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$205.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1672206

API family

topiramate

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NS(OC[C@]12[C@](OC(C)(C)O2)([H])[C@@]3([H])[C@@](OC(C)(C)O3)([H])CO1)(=O)=O

InChI

1S/C12H21NO8S/c1-10(2)18-7-5-16-12(6-17-22(13,14)15)9(8(7)19-10)20-11(3,4)21-12/h7-9H,5-6H2,1-4H3,(H2,13,14,15)/t7-,8-,9+,12+/m1/s1

InChI key

KJADKKWYZYXHBB-XBWDGYHZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Topiramate, a sulfamate-substituted derivative of the monosaccharide D-fructose [1], is a medication, which is widely used for the treatment of epilepsy.[1][2]

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Topiramate may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations[1][2] using high-performance liquid chromatography technique[1] and spectrophotometric technique.[2]

Biochem/physiol Actions

Kainate GluR5 receptor antagonist; anticonvulsant.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2801 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a new method for determination of topiramate in bulk and pharmaceutical formulation using high performance liquid chromatography-UV detection after pre-column derivatization
Majnooni BM, et al.
Journal of Reports in Pharmaceutical Sciences, 3(2), 179-183 (2014)
Optimized and validated flow-injection spectrophotometric analysis of topiramate, piracetam and levetiracetam in pharmaceutical formulations
Hadad G, et al.
Acta pharmaceutica, 61(4), 377-389 (2011)

Questions

  1. What is the solubility of the product Topiramate PHR1600 (CAS 97240-79-4)?

    1 answer
    1. Specific solubility information for the product is not available. The product is expected to be generally soluble in DMSO (40mg/mL), ethanol (20-60mg/mL), and DMF (~25mg/mL).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service