Skip to Content
Merck
All Photos(1)

Key Documents

PHR1170

Supelco

Residual Solvent - Methanol

Pharmaceutical Secondary Standard; Certified Reference Material

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 1.2 ML
$194.00

$194.00


Available to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
3 X 1.2 ML
$194.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$194.00


Available to ship on10 April 2025Details


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

suitable for USP 467
traceable to USP 1601623

CofA

current certificate can be downloaded

application(s)

environmental
pharmaceutical (small molecule)

format

single component solution

storage temp.

2-30°C

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Residual Solvent - Methanol is a CRM and a Secondary Pharmaceutical Reference Standard, traceable and qualified against the corresponding Pharmacopeial standards. Sigma-Aldrich offers Certified Reference Materials (CRMs) for the analysis of Residual Solvents. A comprehensive Certificate of Analysis offers details of the methodology and results of the qualification.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7144 in the slot below. This is an example certificate only and may not be the lot that you receive.

Analyte

Description

    Methanol

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - Solvent

Flash Point(C)

87 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service