Skip to Content
Merck
All Photos(1)

Key Documents

PHR1064

Supelco

Residual Solvents Mixture - Class IIA

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Class 2A Solvent Standard Mix

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 1.2 ML
$203.00

$203.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
3 X 1.2 ML
$203.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$203.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

suitable for USP 467
traceable to USP 1601281

CofA

current certificate can be downloaded

application(s)

environmental
pharmaceutical (small molecule)

format

multi-component solution

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Residual Solvents Mixture - Class IIA are certified reference materials (CRMs) for the analysis of residual solvents. The residual solvents can be found as mixtures, as single-analyte standards, as well as neat CRMs. These CRMs are secondary pharmaceutical reference standards and are traceable and qualified against the corresponding Pharmacopeial standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB7797 in the slot below. This is an example certificate only and may not be the lot that you receive.

Analyte

Description

    Acetonitrile 2 mg/mL
    Chlorobenzene 2 mg/mL
    Cumene 0.3 mg/mL
    Cyclohexane 14 mg/mL
    cis-1,2-Dichloroethene 4 mg/mL
    trans-1,2-Dichloroethene 4 mg/mL
    Methylene Chloride 3 mg/mL
    1,4-Dioxane 2 mg/mL
    Methanol 15 mg/mL
    Methylcyclohexane 5 mg/mL
    Tetrahydrofuran 4 mg/mL
    Toluene 4 mg/mL
    Ethylbenzene 2 mg/mL
    p-Xylene 1 mg/mL
    m-Xylene 1 mg/mL
    o-Xylene 7 mg/mL
    See All (16)

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seung Baek Lee et al.
Nature cell biology, 21(8), 940-951 (2019-07-31)
The receptor-interacting serine/threonine-protein kinases RIPK1 and RIPK3 play important roles in necroptosis that are closely linked to the inflammatory response. Although the activation of necroptosis is well characterized, the mechanism that tunes down necroptosis is largely unknown. Here we find

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service