Skip to Content
Merck
All Photos(1)

Key Documents

72940

Sigma-Aldrich

Ethyl 4-nitrobenzoate

≥98.0% (GC)

Synonym(s):

4-(Ethoxycarbonyl)nitrobenzene, 4-Ethoxycarbonyl-1-nitrobenzene, 4-Nitrobenzoic acid ethyl ester, Ethyl p-nitrobenzoate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$83.30

$83.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$83.30

About This Item

Linear Formula:
O2NC6H4CO2C2H5
CAS Number:
Molecular Weight:
195.17
Beilstein:
1912879
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$83.30


Check Cart for Availability

Request a Bulk Order

Assay

≥98.0% (GC)

form

crystals

mp

55-59 °C (lit.)

functional group

ester
nitro

SMILES string

CCOC(=O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C9H9NO4/c1-2-14-9(11)7-3-5-8(6-4-7)10(12)13/h3-6H,2H2,1H3

InChI key

PHWSCBWNPZDYRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neil R Hartman et al.
Regulatory toxicology and pharmacology : RTP, 70(1), 182-188 (2014-07-11)
The clinical use of local anesthetic products to anesthetize mucous membranes has been associated with methemoglobinemia (MetHba), a serious condition in which the blood has reduced capacity to carry oxygen. An evaluation of spontaneous adverse event reporting of MetHba submitted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service